2,1,3-Benzothiadiazol-5-sulfonylchlorid

Lieferant: CHEMPUR

CC09203.250
CHMPCC09203.250EA 0 EUR
CHMPCC09203.250
2,1,3-Benzothiadiazol-5-sulfonylchlorid
2,1,3-Benzothiadiazol-5-sulfonylchlorid
Formel: C₆H₃ClN₂O₂S₂
MDL: MFCD02681993
CAS-Nummer: 337508-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR