2-Benzo[b]thiophencarbonsäure

Lieferant: CHEMPUR

CC12201.250 CC12201DA
CHMPCC12201.250EA 0 EUR
CHMPCC12201.250 MAYBCC12201DA
2-Benzo[b]thiophencarbonsäure
2-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
Molecular Weight: 178,22 g/mol
MDL: MFCD00051636
CAS-Nummer: 6314-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR