2-Benzo[b]thiophenboronsäure

Lieferant: CHEMPUR

CC12212.250
CHMPCC12212.250EA 0 EUR
CHMPCC12212.250
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR