Benzo[b]thiophen-3-carbaldehyd

Lieferant: CHEMPUR

Synonyme: Benzo[b]thiophene-3-carbaldehyde

CC12304.250
CHMPCC12304.250EA 0 EUR
CHMPCC12304.250
Benzo[b]thiophen-3-carbaldehyd
Benzo[b]thiophen-3-carbaldehyd
Formel: C₉H₆OS
Molecular Weight: 162.2083 g/mol
MDL: MFCD00052376
CAS-Nummer: 5381-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR