3-Benzo[b]thiophenboronsäure
Lieferant: CHEMPUR
Synonyme:
Benzothiophene-3-boronic acid, Benzo[b]thiophen-3-ylboronic acid
CC12312.250
CHMPCC12312.250EA
0
EUR
CHMPCC12312.250
3-Benzo[b]thiophenboronsäure
3-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S Molecular Weight: 178.02 g/mol |
MDL:
MFCD01114669 CAS-Nummer: 113893-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...