3-Indolcarbaldehyd

Lieferant: CHEMPUR

Synonyme: Indol-3-carboxaldehyd

CC12704.250 CC12704.1
CHMPCC12704.250EA 0 EUR
CHMPCC12704.250 MAYBCC12704.1
3-Indolcarbaldehyd
3-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Schmelzpunkt: 192…195 °C
Dichte: 1.34 g/cm³ (20 °C)
MDL: MFCD00005622
CAS-Nummer: 487-89-8
EINECS: 207-665-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR