3-Thiophenboronsäure

Lieferant: CHEMPUR

Synonyme: 3-Thienylboronsäure, Thiophen-3-boronsäure

CC13112.250 CC13112DA
CHMPCC13112.250EA 0 EUR
CHMPCC13112.250 MAYBCC13112DA
3-Thiophenboronsäure
3-Thiopheneboronic acid
Formel: C₄H₅BO₂S
Molecular Weight: 127.96 g/mol
Schmelzpunkt: 162…164 °C
MDL: MFCD00151851
CAS-Nummer: 6165-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR