Ethoxymethylenmalononitril

Lieferant: CHEMPUR

CD04255.250 CD04255.1
CHMPCD04255.250EA 0 EUR
CHMPCD04255.250 MAYBCD04255.1
Ethoxymethylenmalononitril
Ethoxymethylenmalononitril
Formel: C₆H₆N₂O
Molecular Weight: 122.13 g/mol
MDL: MFCD00001854
CAS-Nummer: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR