2-Chlor-6-fluorbenzylamin
Lieferant: CHEMPUR
CD05338.250
CD05338.1
CHMPCD05338.250EA
0
EUR
CHMPCD05338.250
MAYBCD05338.1
2-Chlor-6-fluorbenzylamin
2-Chlor-6-fluorbenzylamin
Formel:
C₇H₇ClFN Molecular Weight: 159.59 g/mol Siedepunkt: 99…100 °C (1013 hPa) |
MDL:
MFCD00042458 CAS-Nummer: 15205-15-9 EINECS: 239-259-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...