4,5-Dibrom-2-thiophensulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
4,5-Dibrom-2-thiophensulfonsäurechlorid, 4,5-Dibromthiophen-2-sulfonylchlorid
CD08466.0001
CD08466.250
MAYBCD08466.0001EA
0
EUR
MAYBCD08466.0001
CHMPCD08466.250
4,5-Dibrom-2-thiophensulfonylchlorid
4,5-Dibrom-2-thiophensulfonylchlorid
Formel:
C₄HBr₂ClO₂S₂ Molecular Weight: 340.44 g/mol Schmelzpunkt: 80 |
MDL:
MFCD00041439 CAS-Nummer: 81606-31-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...