3-Chlor-2-fluorbenzamid
Lieferant: CHEMPUR
Synonyme:
3-Chlor-2-fluorbenzoesäureamid
CD11825.250
CD11825.1
CHMPCD11825.250EA
0
EUR
CHMPCD11825.250
MAYBCD11825.1
3-Chlor-2-fluorbenzamid
3-Chlor-2-fluorbenzamid
Formel:
C₇H₅ClFNO Molecular Weight: 173.57 g/mol |
MDL:
MFCD00042205 CAS-Nummer: 104326-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...