4-(Trifluormethylthio)toluol

Lieferant: CHEMPUR

CF3.25 CF3.5 011227-5G 011227-25G 011227-1G
CHMPCF3.25EA 0 EUR
CHMPCF3.25 CHMPCF3.5 FLUO011227-5G FLUO011227-25G FLUO011227-1G
4-(Trifluormethylthio)toluol
4-(Trifluormethylthio)toluol
Formel: C₈H₇F₃S
MDL: MFCD00129190
CAS-Nummer: 352-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR