4'-Ethoxyacetophenon

Lieferant: CHEMPUR

DP00432.250 DP00432.1
CHMPDP00432.250EA 0 EUR
CHMPDP00432.250 MAYBDP00432.1
4'-Ethoxyacetophenon
4'-Ethoxyacetophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Siedepunkt: 83 °C (0.04 mmHg)
Schmelzpunkt: 39
MDL: MFCD00009095
CAS-Nummer: 1676-63-7
EINECS: 216-825-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR