2-Pyridincarbaldehydoxim

Lieferant: CHEMPUR

Synonyme: 2-Pyridinecarbaldehyde oxime, 2-Pyridinealdoxime

DP01482.250 DP01482.1
CHMPDP01482.250EA 0 EUR
CHMPDP01482.250 MAYBDP01482.1
2-Pyridincarbaldehydoxim
2-Pyridincarbaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122.13 g/mol
MDL: MFCD00006354
CAS-Nummer: 873-69-8
EINECS: 212-849-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR