3,4-Dimethylbenzoesäure
Lieferant: CHEMPUR
DP01901.250
DP01901.1
CHMPDP01901.250EA
0
EUR
CHMPDP01901.250
MAYBDP01901.1
3,4-Dimethylbenzoesäure
3,4-Dimethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 g/mol Siedepunkt: 295 Schmelzpunkt: 163…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002524 CAS-Nummer: 619-04-5 EINECS: 210-576-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...