4-Bromisophthalsäure

Lieferant: CHEMPUR

FL1005.25
CHMPFL1005.25EA 0 EUR
CHMPFL1005.25
4-Bromisophthalsäure
4-Bromisophthalsäure
Formel: C₈H₅BrO₄
Molecular Weight: 245,03 g/mol
Schmelzpunkt: 303…304 °C
MDL: MFCD00002404
CAS-Nummer: 6939-93-1
EINECS: 230-078-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR