7-Methyladenin

Lieferant: CHEMPUR

Synonyme: 6-Amino-7-methylpurin

FL10215.POR
CHMPFL10215.POREA 0 EUR
CHMPFL10215.POR
7-Methyladenin
7-Methyladenin
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
MDL: MFCD00127790
CAS-Nummer: 935-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR