10-Hydroxydecansäure

Lieferant: CHEMPUR

FL10221.POR
CHMPFL10221.POREA 0 EUR
CHMPFL10221.POR
10-Hydroxydecansäure
10-Hydroxydecanoic acid
Formel: C₁₀H₂₀O₃
Molecular Weight: 188,26 g/mol
MDL: MFCD00010510
CAS-Nummer: 1679-53-4
EINECS: 216-848-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR