15-Hydroxypentadecansäure

Lieferant: CHEMPUR

FL10223.POR
CHMPFL10223.POREA 0 EUR
CHMPFL10223.POR
15-Hydroxypentadecansäure
15-Hydroxypentadecanoic acid
Formel: C₁₅H₃₀O₃
MDL: MFCD00039535
CAS-Nummer: 4617-33-8
EINECS: 225-026-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR