2-Iod-1-phenylethan
Lieferant: CHEMPUR
Synonyme:
2-Phenylethyliodid
FL1087.POR
CHMPFL1087.POREA
0
EUR
CHMPFL1087.POR
2-Iod-1-phenylethan
2-Iod-1-phenylethan
Formel:
C₆H₅CH₂CH₂I Molecular Weight: 232,06 g/mol Siedepunkt: 125 °C (18 mmHg) Schmelzpunkt: –2 °C Dichte: 1,6323 g/cm³ (20 °C) |
MDL:
MFCD00039408 CAS-Nummer: 17376-04-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...