3-Acetylphenylboronsäure

Lieferant: CHEMPUR

Synonyme: (3-Acetylphenyl)metaborsäure, Dihydroxy(3-acetylphenyl)boran, 3-Acetylbenzolboronsäure

FL10976.POR
CHMPFL10976.POREA 0 EUR
CHMPFL10976.POR
3-Acetylphenylboronsäure
3-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
MDL: MFCD01074678
CAS-Nummer: 204841-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR