2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan

Lieferant: FLUOROCHEM

Synonyme: Isopropyl pinacolyl borate, 4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

010803-100G FL11000.POR 010803-25G
FLUO010803-100GEA 194 EUR
FLUO010803-100G CHMPFL11000.POR FLUO010803-25G
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
Formel: C₉H₁₉BO₃
Molecular Weight: 186.06 g/mol
MDL: MFCD00192241
CAS-Nummer: 61676-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR