2,4-Dibenzyloxy-5-pyrimidinboronsäure

Lieferant: CHEMPUR

Synonyme: 2,4-Bis(benzyloxy)pyrimidine-5-boronic acid, 2,4-Bis(benzyloxy)pyrimidin-5-ylboronic acid

FL11016.POR
CHMPFL11016.POREA 0 EUR
CHMPFL11016.POR
2,4-Dibenzyloxy-5-pyrimidinboronsäure
2,4-Dibenzyloxy-5-pyrimidinboronsäure
Formel: C₁₈H₁₇BN₂O₄
Molecular Weight: 336,150 g/mol
MDL: MFCD01318938
CAS-Nummer: 70523-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR