3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion

Lieferant: CHEMPUR
Gefahr

Synonyme: 3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione

FL11043.POR
CHMPFL11043.POREA 0 EUR
CHMPFL11043.POR
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion
Formel: C₁₉H₃₄O₃Sn
Molecular Weight: 429,17 g/mol
MDL: MFCD01319011
CAS-Nummer: 129034-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR