4-(Methylsulfonyl)phenylboronsäure

Lieferant: CHEMPUR

Synonyme: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

FL11044.POR
CHMPFL11044.POREA 0 EUR
CHMPFL11044.POR
4-(Methylsulfonyl)phenylboronsäure
4-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200.02 g/mol
MDL: MFCD01630820
CAS-Nummer: 149104-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR