2,4,6-Trichlorphenylboronsäure

Lieferant: CHEMPUR

FL11063.POR
CHMPFL11063.POREA 0 EUR
CHMPFL11063.POR
2,4,6-Trichlorphenylboronsäure
2,4,6-Trichlorphenylboronsäure
Formel: C₆H₄BCl₃O₂
Molecular Weight: 225,26 g/mol
MDL: MFCD01074621
CAS-Nummer: 73852-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR