2,5-Dimethoxybenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 2,5-Dimethoxybenzol-1-sulfonylchlorid, 2,5-Dimethoxybenzolsulfochlorid, 2,5-Dimethoxybenzolsulfonsäurechlorid

FL11154.POR
CHMPFL11154.POREA 0 EUR
CHMPFL11154.POR
2,5-Dimethoxybenzolsulfonylchlorid
2,5-Dimethoxybenzolsulfonylchlorid
Formel: C₈H₉ClO₄S
Molecular Weight: 236,68 g/mol
Schmelzpunkt: 119…121 °C
MDL: MFCD00024872
CAS-Nummer: 1483-28-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR