3,5-Dichloranthranilsäure
Lieferant: CHEMPUR
Synonyme:
2-Amino-3,5-dichlorobenzoic acid
FL11350.POR
CHMPFL11350.POREA
0
EUR
CHMPFL11350.POR
3,5-Dichloranthranilsäure
3,5-Dichloranthranilsäure
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,03 g/mol Siedepunkt: 344 °C (1013 hPa) Schmelzpunkt: 226…229 °C |
MDL:
MFCD00017088 CAS-Nummer: 2789-92-6 EINECS: 220-520-3 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...