4-Chlorzimtsäure

Lieferant: CHEMPUR

Synonyme: 4-Chlorcinnamsäure, 3-(p-Chlorphenyl)propensäure, 3-(4-Chlorphenyl)propensäure

FL1144.100
CHMPFL1144.100EA 0 EUR
CHMPFL1144.100
4-Chlorzimtsäure
4-Chlorzimtsäure
Formel: C₉H₇ClO₂
Molecular Weight: 182,61 g/mol
Siedepunkt: 311 °C (1013 hPa)
Schmelzpunkt: 252…254 °C
MDL: MFCD00004396
CAS-Nummer: 1615-02-7
EINECS: 216-564-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR