5-Bromuracil

Lieferant: CHEMPUR

Synonyme: Bromuracil, 5-Brom-2,4-dihydroxypyrimidin

FL11596.1
CHMPFL11596.1EA 0 EUR
CHMPFL11596.1
5-Bromuracil
5-Bromuracil
Formel: C₄H₃BrN₂O₂
Molecular Weight: 190,98 g/mol
Siedepunkt: 384 °C (1013 hPa)
Schmelzpunkt: 165 °C
MDL: MFCD00006017
CAS-Nummer: 51-20-7
EINECS: 200-084-0
Merck Index: 13,01425

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR