5-Hydroxyanthranilsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
5-Hydroxyanthranilic acid
FL11772.POR
CHMPFL11772.POREA
0
EUR
CHMPFL11772.POR
5-Hydroxyanthranilsäure
5-Hydroxyanthranilsäure
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 255…265 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007870 CAS-Nummer: 394-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...