3-Fluor-2-(trifluormethyl)benzaldehyd

Lieferant: CHEMPUR

FL11915.POR
CHMPFL11915.POREA 0 EUR
CHMPFL11915.POR
3-Fluor-2-(trifluormethyl)benzaldehyd
3-Fluoro-2-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
MDL: MFCD04116354
CAS-Nummer: 924817-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR