Methylmercaptoacetat

Lieferant: CHEMPUR

Synonyme: Methyl thioglycolate

FL11917.POR
CHMPFL11917.POREA 0 EUR
CHMPFL11917.POR
Methylmercaptoacetat
Methylmercaptoacetat
Formel: C₃H₆O₂S
Molecular Weight: 106.15 g/mol
MDL: MFCD00004873
CAS-Nummer: 2365-48-2
EINECS: 219-121-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR