6-Chlor-3-formylchromon

Lieferant: CHEMPUR

Synonyme: 6-Chloro-4-oxo-4H-chromene-3-carbaldehyde

FL12003.POR 012003-25G
CHMPFL12003.POREA 0 EUR
CHMPFL12003.POR FLUO012003-25G
6-Chlor-3-formylchromon
6-Chlor-3-formylchromon
Formel: C₁₀H₅ClO₃
Molecular Weight: 208.597 g/mol
MDL: MFCD00139138
CAS-Nummer: 42248-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR