6-Chlor-3-formyl-7-methylchromon

Lieferant: CHEMPUR

FL12005.POR 012005-5G
CHMPFL12005.POREA 0 EUR
CHMPFL12005.POR FLUO012005-5G
6-Chlor-3-formyl-7-methylchromon
6-Chlor-3-formyl-7-methylchromon
Formel: C₁₁H₇ClO₃
MDL: MFCD00191919
CAS-Nummer: 64481-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR