6,8-Dichlor-3-formylchromon

Lieferant: CHEMPUR

FL12006.POR 012006-25G
CHMPFL12006.POREA 0 EUR
CHMPFL12006.POR FLUO012006-25G
6,8-Dichlor-3-formylchromon
6,8-Dichlor-3-formylchromon
Formel: C₁₀H₄Cl₂O₃
MDL: MFCD00051510
CAS-Nummer: 64481-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR