6-Ethyl-3-formylchromon

Lieferant: CHEMPUR

FL12007.POR 012007-5G
CHMPFL12007.POREA 0 EUR
CHMPFL12007.POR FLUO012007-5G
6-Ethyl-3-formylchromon
6-Ethyl-3-formylchromon
Formel: C₁₂H₁₀O₃
MDL: MFCD00192155
CAS-Nummer: 42059-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR