5,7-Dimethyl-3-formylchromon

Lieferant: CHEMPUR

FL12012.POR 012012-5G
CHMPFL12012.POREA 0 EUR
CHMPFL12012.POR FLUO012012-5G
5,7-Dimethyl-3-formylchromon
5,7-Dimethyl-3-formylchromon
Formel: C₁₂H₁₀O₃
MDL: MFCD00051509
CAS-Nummer: 62484-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR