6,7-Dimethyl-3-formylchromon

Lieferant: CHEMPUR

FL12013.POR
CHMPFL12013.POREA 0 EUR
CHMPFL12013.POR
6,7-Dimethyl-3-formylchromon
6,7-Dimethyl-3-formylchromon
Formel: C₁₂H₁₀O₃
MDL: MFCD00192181
CAS-Nummer: 57803-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR