3-Cyan-6-methylchromon

Lieferant: CHEMPUR

FL12016.POR
CHMPFL12016.POREA 0 EUR
CHMPFL12016.POR
3-Cyan-6-methylchromon
3-Cyan-6-methylchromon
Formel: C₁₁H₇NO₂
MDL: MFCD00191962
CAS-Nummer: 50743-18-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR