6-Chlor-3-cyanchromon

Lieferant: CHEMPUR

Synonyme: 6-Chlorochromone-3-carbonitrile

FL12017.POR
CHMPFL12017.POREA 0 EUR
CHMPFL12017.POR
6-Chlor-3-cyanchromon
6-Chlor-3-cyanchromon
Formel: C₁₀H₄ClNO₂
Molecular Weight: 205,60 g/mol
MDL: MFCD00191905
CAS-Nummer: 50743-20-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR