2-Amino-6-chlor-3-formylchromon

Lieferant: CHEMPUR

FL12027.POR
CHMPFL12027.POREA 0 EUR
CHMPFL12027.POR
2-Amino-6-chlor-3-formylchromon
2-Amino-6-chlor-3-formylchromon
Formel: C₁₀H₆ClNO₃
MDL: MFCD00139140
CAS-Nummer: 68301-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR