2-Amino-6-brom-3-formylchromon

Lieferant: CHEMPUR

FL12028.POR
CHMPFL12028.POREA 0 EUR
CHMPFL12028.POR
2-Amino-6-brom-3-formylchromon
2-Amino-6-brom-3-formylchromon
Formel: C₁₀H₆BrNO₃
CAS-Nummer: 73262-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR