2-Amino-6-ethyl-3-formylchromon

Lieferant: CHEMPUR

FL12029.POR
CHMPFL12029.POREA 0 EUR
CHMPFL12029.POR
2-Amino-6-ethyl-3-formylchromon
2-Amino-6-ethyl-3-formylchromon
Formel: C₁₂H₁₁NO₃
CAS-Nummer: 68301-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR