2-Amino-3-formyl-6,7-dimethylchromon

Lieferant: CHEMPUR

FL12032.POR
CHMPFL12032.POREA 0 EUR
CHMPFL12032.POR
2-Amino-3-formyl-6,7-dimethylchromon
2-Amino-3-formyl-6,7-dimethylchromon
Formel: C₁₂H₁₁NO₃
MDL: MFCD00191736
CAS-Nummer: 94978-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR