6-Chlorchromon-3-carbonsäure

Lieferant: CHEMPUR

FL12043.POR
CHMPFL12043.POREA 0 EUR
CHMPFL12043.POR
6-Chlorchromon-3-carbonsäure
6-Chlorchromon-3-carbonsäure
Formel: C₁₀H₅ClO₄
CAS-Nummer: 51085-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR