6,8-Dichlor-3-formylchromon

Lieferant: CHEMPUR

Synonyme: 6,8-Dichloro-3-formylchromone, 6,8-Dichlorochromone-3-carboxaldehyde, 6,8-Dichloro-4-oxo-4H-chromene-3-carbaldehyde, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

FL12063.POR
CHMPFL12063.POREA 0 EUR
CHMPFL12063.POR
6,8-Dichlor-3-formylchromon
6,8-Dichlor-3-formylchromon
Formel: C₁₀H₄Cl₂O₃
Molecular Weight: 243,0430 g/mol
MDL: MFCD00051510
CAS-Nummer: 64481-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR