5'-Ethyl-2'-hydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 5-Ethyl-2-hydroxyacetophenone

FL12073.POR 012073-25G
CHMPFL12073.POREA 0 EUR
CHMPFL12073.POR FLUO012073-25G
5'-Ethyl-2'-hydroxyacetophenon
5'-Ethyl-2'-hydroxyacetophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2011 g/mol
MDL: MFCD00053308
CAS-Nummer: 24539-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR