2'-Hydroxy-5'-isopropyl-3'-nitroacetophenon

Lieferant: CHEMPUR

FL12084.POR
CHMPFL12084.POREA 0 EUR
CHMPFL12084.POR
2'-Hydroxy-5'-isopropyl-3'-nitroacetophenon
2'-Hydroxy-5'-isopropyl-3'-nitroacetophenon
Formel: C₁₁H₁₃NO₄
CAS-Nummer: 288401-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR