5'-Chlor-2'-hydroxy-4'-methylpropiophenon

Lieferant: CHEMPUR

FL12089.POR
CHMPFL12089.POREA 0 EUR
CHMPFL12089.POR
5'-Chlor-2'-hydroxy-4'-methylpropiophenon
5'-Chlor-2'-hydroxy-4'-methylpropiophenon
Formel: C₁₀H₁₁ClO₂
MDL: MFCD02093255
CAS-Nummer: 22362-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR